missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FreeStyle™ MAX 293 Expression System

Product Code. 10746033
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10746033

Brand: Invitrogen™ K900010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapid and high-yield mammalian protein production

FreeStyle MAX Reagent is a novel cationic lipid-based reagent designed to transfect FreeStyle cells with high efficiency. The FreeStyle MAX Expression System combines GIBCO™ FreeStyle Media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293 cells. The only equipment required to use the FreeStyle MAX System is a CO2 incubator and a platform shaker with shake flasks or a spinner culture

FreeStyle MAX System provides:

  • Easy, rapid protocol for transient transfection of CHO and 293 cells in suspension cultures
  • Functional proteins (up to milligram quantities) with all post-translational modifications produced in one week
  • Simplified downstream purification of secreted proteins with FreeStyle serum-free medium
  • Straightforward protocol for production scale-up
  • Replace time-consuming stable cell line generation with large-scale functional protein production in one week

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Expression, Mammalian Protein Production, Plasmid Transfection, Protein Expression, Proteins, Expression, Isolation and Analysis, Transfection, Transient Protein Production and High-Throughput Screening

TRUSTED_SUSTAINABILITY

Specifications

Cell Line FreeStyle™ 293-F Cells
Classification Animal Origin-Free, Serum-Free
Content And Storage Store FreeStyle cells in liquid nitrogen. Store FreeStyle MAX Reagent at 2–8°C. Store FreeStyle CHO Expression Medium, FreeStyle 293 Expression Medium, and OptiPRO SFM at 2–8°C in the dark. Store the pCMV SPORT at -20°C. Stability is guaranteed for 6 months under the recommended storage conditions. Please refer to the manual for additional storage information.
Delivery Type Transfection
Culture Type Suspension Cell Culture
Cell Type 293-derived Cells
Transfection Technique Lipid-based Transfection, Transient Transfection
Expression Mechanism Cell-Based Expression
Expression System Mammalian
For Use With (Application) Protein Expression
Key Functions Protein Production
Product Line Freestyle
Product Type Expression System
Quantity 1 Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.