missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ β-Gal Staining Kit

Product Code. 10247282
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10247282 1 Kit 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10247282 Supplier Invitrogen™ Supplier No. K146501
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Sufficient reagents to stain fifty 60mM plates. The kit includes 10X PBS (phosphate buffered saline), X-gal, staining solutions A, B, and C, 10X fixative solution, and 10μg of pcDNA 3.1/His/lacZ control vector.

Determine percentage of transfected cells expressing lacZ

  • lacZ is bacterial gene often used as reporter construct in eukaryotic transfection experiments
  • Gene product of lacZ, β-galactosidase, is resistant to proteolysis in cellular lysates, and its activity is easily assayed
  • β-galactosidase catalyzes hydrolysis of X-gal, producing a blue color that can be easily visualized under microscope
  • Colorimetric detection

Beta-Galactosidase Assays/Enzyme and Protein Activity Assays/Proteins, Expression, Isolation and Analysis/Reporter Assays

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The β-Gal Staining Kit contains sufficient reagents to stain fifty 60 mm plates. The kit includes 10X PBS (phosphate buffered saline), X-gal, staining solutions A, B, and C, 10X fixative solution, and 10 μg of pcDNA™ 3.1/His/lacZ control vector. Store the 10X PBS at room temperature. All other components should be stored at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Assay β-Gal (lacZ) Assay
Detection Method Colorimetric
Product Type Beta-Gal Staining Kit
Substrate X-Gal
Quantity 1 Kit
Target Beta-Galactosidase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.