missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pAd/CMV/V5-DEST™ Gateway™ Vector Kit

Product Code. 11540836
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11540836 6 μg 6µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11540836 Supplier Invitrogen™ Supplier No. V49320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

pAd-CMV/V5-DEST Vector 150ng/μL in TEBuffer, pH 8.0. 40μL; pAd/CMV/V5-GW/lacZ control plasmid 1μg/μL in TE Buffer, pH 8.0, 10μL.

For easy recombination-based cloning and adenoviral-based, transient expression of a target gene in dividing and nondividing mammalian cells

  • The vector allows generation of an adenovirus containing the target gene where constitutive, high-level expression is driven by the CMV promoter
  • High efficiency and rapid recombination cloning
  • Produces high titer adenoviral stocks
  • Efficient delivery of the gene to dividing and nondividing mammalian cells in vitro or in vivo
  • Produces replication-incompetent virus for enhanced biosafety of the system
  • Amenable for use in high-throughput applications
  • CMV promoter for high-level constitutive expression of gene of interest
  • Human Ad5 sequences (ΔE3) and Viral Inverted Terminal Repeats (ITRs) for packaging of the expression construct into virions
  • Herpes Simplex Virus thymidine kinase (TK) polyadenylation sequence for efficient transcription termination and polyadenylation of mRNA
  • V5 epitope for detection of recombinant protein
  • Ampicillin selection marker

Antibody and Cell Culture Production, Bioproduction, Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Adenoviral
Promoter CMV
Product Type Mammalian Expression Vector
Content And Storage • pAd-CMV⁄V5-DEST™ Vector 150 ng⁄μl in TEBuffer, pH 8.0. 40 μl (Store at -20°C)
• pAd⁄CMV⁄V5-GW⁄lacZ control plasmid 1 μg⁄μl in TE Buffer, pH 8.0. 10 μl (Store at -20°C)
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 6 μg
Vector pDEST
Product Line Gateway, ViraPower
For Use With (Application) Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.