missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pcDNA™3.2-DEST Mammalian Expression Vector
Click to view available options
Quantity:
6 μg
Unit Size:
1 set
Description
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
- Cytomegalovirus (CMV) promoter for high-level expression
- attR sites for Gateway cloning, enabling recombination with attL-flanked fragments
- C-terminal V5 tag for easy detection
- Neomycin resistance gene for stable selection
- Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
- Gateway Cloning
Antibody and Cell Culture Production,Bioproduction,Cloning,Constitutive Expression,Gateway Cloning,Mammalian Expression.Protein Expression,Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Mammalian Expression Vector |
| Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
| Content And Storage | All destination vectors are provided lyophilized and supercoiled. |
| Protein Tag | V5 Epitope Tag |
| Cloning Method | Gateway |
| Quantity | 6 μg |
| Vector | pDEST, pcDNA |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction