missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ pcDNA™-DEST47 Vector

Product Code. 11538696
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538696 6 μg 6µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538696 Supplier Invitrogen™ Supplier No. 12281010
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

All destination vectors are provided lyophilized and supercoiled.

Requires

Gateway entry clone, appropriate Gateway LR Clonase™ enzyme mix, and reaction buffer.

Designed for high-level, constitutive expression in variety of mammalian cell lines

  • C-terminal Cycle 3 GFP tag for rapid detection of recombinant protein
  • Cytomegalovirus (CMV) promoter for high-level expression
  • attR sites for Gateway cloning, enabling recombination with attL-flanked fragments
  • Neomycin resistance gene for stable selection
  • Ampicillin resistance gene and pUC origin for selection and maintenance in E. Coli

Cloning,Constitutive Expression,Gateway Cloning,Mammalian Expression,Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter CMV
Product Type Gateway System Destination Expression Vector
Reporter Gene GFP (Cycle 3)
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Protein Tag GFP (Cycle 3)
Cloning Method Gateway
Quantity 6 μg
Vector pDEST, pcDNA
Product Line Gateway, pDEST
For Use With (Application) Constitutive Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.