Learn More
Invitrogen™ Gateway™ pDEST™10 Vector
Product Code. 11598666
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
Product Code. Quantity unitSize
11598666 6 μg 6µg
1 options
Product Code. 11598666 Supplier Invitrogen™ Supplier No. 11806015
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins
  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Baculovirus Production and Titering,Cloning,Gateway Cloning,Insect Expression,Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter Polyhedrin
Product Type Gateway System Destination Expression Vector
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Cleavage TEV Protease Recognition Site
Protein Tag His Tag (6x)
Cloning Method Gateway
Quantity 6 μg
Vector pDEST, Gateway Baculovirus Vectors
Product Line Gateway