missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ pDEST™24 Vector
Description
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Bacterial Expression, Cloning, Gateway Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Constitutive or Inducible System | Inducible |
| Promoter | T7 |
| Product Type | Gateway System Destination Expression Vector |
| Selection Agent (Eukaryotic) | None |
| Content And Storage | All destination vectors are provided lyophilized and supercoiled. |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
| Protein Tag | GST Tag |
| Cloning Method | Gateway |
| Quantity | 6 μg |
| Vector | pDEST, Gateway T7 Vectors |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?