missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti6.2-GW/EmGFP Expression Control Vector

Product Code. 10153752
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10153752

Brand: Invitrogen™ V36920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

pLenti6.2-GW/EmGFP Vector: 20μg of control vector is supplied in solution as 40μL of 0.5μg/μL in 10mM Tris–HCl, 1mM EDTA, pH 8.0.

Designed as a positive control to enable the detection of EmGFP fluorescence following transfection in 293FT cells

  • Also a titer control to produce an EmGFP-expressing lentivirus stock and a transduction control following transduction in both dividing and nondividing mammalian cells
  • This is not a cloning vector
  • Human cytomegalovirus (CMV) immediate early promoter to control high-level expression of the EmGFP
  • PKG Promoter for expression of Blasticidin selection marker

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Cloning, Constitutive Expression, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays, Viral Gene Delivery

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Promoter CMV
Product Type Lentiviral Expression Vector
Reporter Gene GFP (EmGFP)
Selection Agent (Eukaryotic) Blasticidin
Content And Storage pLenti6.2-GW⁄EmGFP Vector: 20 μg of control vector is supplied in solution as 40 μl of 0.5 μg⁄μl in 10 mM Tris–HCl, 1 mM EDTA, pH 8.0. Store at –20°C.
Selection Marker Promoter PGK Promoter
Protein Tag Untagged
Cloning Method Gateway
Quantity 1 Ea.
Vector pLenti
Product Line Gateway, ViraPower, Vivid Colors
For Use With (Application) Reporter Assays, Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.