missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ pT-Rex™-DEST30 Vector

Product Code. 11508706
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11508706 6 μg 6µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11508706 Supplier Invitrogen™ Supplier No. 12301016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

All destination vectors are provided lyophilized and supercoiled.

Requires

Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.

Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins

  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone

Cloning, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis/Regulated Expression

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Inducible
Delivery Type Transfection
Inducing Agent Tetracycline
Promoter CMV/TO
Product Type Gateway System Destination Expression Vector
Selection Agent (Eukaryotic) Neomycin⁄Kanamycin
Content And Storage All destination vectors are provided lyophilized and supercoiled.
Protein Tag Untagged
Cloning Method Gateway
Quantity 6 μg
Vector pDEST
Product Line Gateway, T-REx
For Use With (Application) Regulated Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.