missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViraPower™ UbC Lentiviral Gateway™ Expression Kit

Product Code. 17115999
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17115999 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17115999 Supplier Invitrogen™ Supplier No. K499000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Includes

pLenti6/UbC/V5-DEST (Box 1): 6μg; pLenti6/UbC/V5-GW/lacZ Control vector (Box 1): 10μg; Stbl3 Chemically Competent E. coli (Box 2): 21 x 50μL; pUC19 control DNA (10pg/μL): 50μL; S.O.C. Medium: 6mL; Support kit (Box 3); ViraPower Packaging Mix (1μg/μL): 195μg; Lipofectamine 2000: 0.75mL; Blasticidin: 50mg; 293 FT cells (3 x 106/mL) (Box 4): 1mL

Facilitates easy recombination-based cloning and lentiviral-based expression of a target gene in dividing and nondividing mammalian cells

  • Contains all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit
  • The pLenti6/UbC/V5-DEST vector has the UbC promoter for driving constitutive but physiological levels of expression of the target gene and the blasticidin selection marker for stable selection in mammalian cells
  • Well-suited to long-term experiments
  • Accurate titer of functional virus
  • Flexible and versatile Gateway recombination cloning technology
  • Vector includes UbC promoter and V5 epitope tag at C terminus

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Promoter UbC
Product Type Lentiviral Expression Kit
Selection Agent (Eukaryotic) Blasticidin
Content And Storage Vector kit (Boxes 1 and 2)
pLenti6⁄UbC⁄V5-DEST™ (Box 1): 6 μg, -20°C
pLenti6⁄UbC⁄V5-GW⁄lacZ Control vector (Box 1): 10 μg, -20°C
One Shot™ Stbl3™ Chemically Competent E. coli (Box 2): 21 x 50 μL, -80°C
pUC19 control DNA (10 pg⁄μL): 50 μL, -80°C
S.O.C. Medium: 6 mL -80°C

Support kit (Box 3)
ViraPower™ Packaging Mix (1 μg⁄μL): 195 μg, -20°C
Lipofectamine™ 2000: 0.75 mL, +4°C
Blasticidin: 50 mg, -20°C
293 FT cells (3 x 106⁄mL) (Box 4): 1 mL, liquid nitrogen
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 1 kit
Vector pLenti
Product Line Gateway, ViraPower
For Use With (Application) Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.