missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneRacer™ Kit with AMV RT and TOPO TA Cloning™ Kit for Sequencing

Product Code. 10728273
Click to view available options
Quantity:
1 Kit
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 10728273

Brand: Invitrogen™ L150001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Advanced RACE (rapid amplification of cDNA ends) technique that improves efficiency of amplifying full-length 5' and 3' cDNA ends

The GeneRacer Kit is available with SuperScriptIII Reverse Transcriptase (RT) for improved amplification of the full-length 5' end from long and complex mRNA. The RNase H portion of SuperScriptIII RT has been mutated to avoid cleaving mRNA during cDNA synthesis. This increases the size and yield of cDNA. SuperScriptIII RT is more thermostable than wild-type RTs. This enables reverse transcription at higher temperatures, relaxing secondary structure of complex templates, and allowing cDNA synthesis to go to completion.

  • Generate cDNA from transcripts at least 10kb in length
  • Obtain the full-length 5' end of rare transcripts at fewer than 30 copies per cell
  • Clone the full-length 5' and 3' ends to construct complete cDNA sequence

  • Ensures that only transcripts containing full-length cDNA ends are amplified (1,2)
  • Advanced protocol starts at RNA level by specifically targeting only 5' capped mRNA
  • In subsequent steps, cap is removed and replaced with GeneRacer RNA Oligo
  • During reverse transcription, GeneRacer RNA Oligo sequence is incorporated into cDNA
  • Only cDNA that is completely reverse transcribed will contain this known sequence
  • 5' RACE PCR is then performed using GeneRacer 5' Primer specific to GeneRacer RNA Oligo sequence and gene-specific primer
  • Result is amplified DNA that contains the full-length 5' cDNA sequence
  • To demonstrate the ability of GeneRacer Kit to capture full-length 5fi cDNA end, 5fi ends of genes with known transcriptional start sites were amplified
  • Starting with total RNA and following GeneRacer protocol, both long transcripts (10 kb) and rare messages present at 0.01%, or 30 copies per cell were amplified

Cloning, PCR and Real-Time PCR, Reverse Transcription, cDNA Libraries and Library Construction

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10
Cloning Method TOPO TA
Content And Storage Each GeneRacer™ Kit contains the GeneRacer™ box, an RT box, S.N.A.P.™ columns, and a TOPO™ Cloning Kit. The GeneRacer™ and the RT box contain sufficient reagents for five cDNA reactions plus one control reaction and primers for 50 PCR reactions. Included in the GeneRacer™ box are the enzymes CIP, TAP, T4 RNA ligase, and their buffers, the GeneRacer™ RNA Oligo (pre-aliquotted and lyophilized), GeneRacer™ Primers and Nested Primers, RNaseOUT™ Recombinant Ribonuclease Inhibitor, phenol/chloroform, mussel glycogen, sterile water, and controls. Store at -20°C. The SuperScript™ III RT box includes SuperScript™ III RT, 5X first-strand buffer, DTT, RNase H, random primers, the GeneRacer™ Oligo dT Primer, and dNTP mix. Store at -20°C. The Cloned AMV RT box includes cloned AMV RT, 5X RT buffer, random primers, the GeneRacer™ Oligo dT Primer, and dNTP mix. Store at -80°C. A separate bag contains ten S.N.A.P.™ gel purification columns for gel-purifying PCR products. Store at room temperature. The 10-reaction TOPO™ Cloning Kit for Sequencing contains two boxes. Store the TOPO™ Cloning box at -20°C. Store the competent E. coli box at -70°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
For Use With (Application) Reverse Transcription
Includes Each GeneRacer Kit contains the GeneRacer box, an RT box, S.N.A.P. columns, and a TOPO Cloning Kit. The GeneRacer and the RT box contain sufficient reagents for five cDNA reactions plus one control reaction and primers for 50 PCR reactions. Included in the GeneRacer box are the enzymes CIP, TAP, T4 RNA ligase, and their buffers, the GeneRacer RNA Oligo (pre-aliquotted and lyophilized), GeneRacer Primers and Nested Primers, RNaseOUT Recombinant Ribonuclease Inhibitor, phenol/chloroform, mussel glycogen, sterile water, and controls. Store at -20°C. The SuperScript III RT box includes SuperScript III RT, 5X first-strand buffer, DTT, RNase H, random primers, the GeneRacer Oligo dT Primer, and dNTP mix. Store at -20°C. The Cloned AMV RT box includes cloned AMV RT, 5X RT buffer, random primers, the GeneRacer Oligo dT Primer, and dNTP mix.
Product Line GeneRacer™, TA Cloning™, TOPO™
Product Type Cloning Kit
Quantity 1 Kit
Vector pCR4-TOPO TA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.