missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GripTite™ 293 MSR Cell Line

Product Code. 10042522
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10042522

Brand: Invitrogen™ R79507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

The GripTite 293 MSR Cell Line is provided as two tubes of a frozen stock of 3 x 106 cells, each in 1mL of 90% complete media and 10% DMSO. Liquid Geneticin Selective Antibiotic (20mL) is supplied as a 50mg/mL solution. Versene (100mL) contains 0.2g/L EDTA 4Na in phosphate-buffered saline. 1X concentration is 0.2g/L, 0.53mM.

A genetically engineered Human Embryonic Kidney (HEK 293) cell line that expresses the human macrophage scavenger receptor

  • Strongly adheres to standard tissue culture plates for dependable results
  • Show the same fast cell growth, high transfection efficiency, and high-level expression characteristics of the parental 293-H cells
  • Unlike most 293 cells, these cells don't wash away during the repeated manipulations of routine tissue culture or plate washing protocols
  • Superior adherence enables reliable reproduction of ligand-binding, enzymatic, or immunofluorescence assay results
  • The GripTite293 MSR cells enhance performance in high-throughput cell-based, as well as standard tissue culture, applications
  • No need for the poly-lysine coated plates typically used in high-throughput analysis: withstand automated plate washing and protocols using liquid handling robots on standard tissue culture plates without significant cell loss

Antibody and Cell Culture Production, Bioproduction, Cell Culture/Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Mammalian Protein Production, Plasmid Transfection, Protein Expression, Proteins, Expression, Isolation and Analysis, RNAi Transfection, Synthetic siRNA Transfection, Transfection, shRNA and miR RNAi Plasmid Transfection

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The GripTite™ 293 MSR Cell Line is provided as two tubes of a frozen stock of 1 x 107 cells, each in 1 ml of 90% complete media and 10% DMSO. Store in liquid nitrogen. Liquid Geneticin™ Selective Antibiotic (20 ml) is supplied as a 50 mg/ml solution. Store at -20°C or 2°C to 8°C. Versene (100 ml) contains 0.2 g/L EDTA 4Na in phosphate-buffered saline. 1X concentration is 0.2 g/L=0.53 mM. Store at 2°C to 8°C. Guaranteed stable for 6 months when properly stored.
Cell Line 293 GripTite™ MSR
Species Human
Concentration 1X
Quantity 1 kit
Product Line GripTite

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.