missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Taq DNA Polymerase, recombinant

Product Code. 10174762
Change view
Click to view available options
No. of Reactions:
100 Reactions
1500 Reactions
500 Reactions
5000 Reactions
Unit Size:
1000 units
500 units
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10174762 100 Reactions Each
10033382 500 Reactions 500 units
11508626 1500 Reactions 1000 units
10063262 5000 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10174762 Supplier Invitrogen™ Supplier No. 10342053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Taq DNA Polymerase (5U/μL); 10X PCR buffer (200mM Tris-HCl pH 8.4, 500mM KCl); Magnesium Chloride (50mM)

Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.

Taq DNA Polymerase, recombinant, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This recombinant enzyme is purified from the cloned Thermus aquaticus DNA polymerase gene expressed in E. coli and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.

Features

  • A choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • Enzyme is licensed and qualified for PCR

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • RT-PCR
  • Sequencing ssDNA
  • Cycle sequencing

Notes

  • One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C.
  • For superior PCR performance, DreamTaq DNA Polymerase is recommended.

Order Info

Shipping Condition: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage Taq DNA Polymerase (5 U/μL), 20 μL
• 10X PCR buffer, 1.5 mL
• 50 mM MgCl2, 1 mL

Store at -10°C to -30°C in a non-frost-free freezer.
Guaranteed stable for 6 months when stored properly.
Format Stand-alone enzyme
GC-Rich PCR Performance Low
PCR Method qPCR, Standard PCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Quantity 100 Units
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
No. of Reactions 100 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.