missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ imMedia™ Growth Medium, agar, kanamycin

Product Code. 10665773
Change view
Click to view available options
Quantity:
20 Ea.
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10665773 20 Ea. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10665773 Supplier Invitrogen™ Supplier No. Q61120

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

imMedia growth medium is provided in a 20-pouch box. Each pouch contains sufficient reagents to prepare 200mL of liquid medium or 8-10 agar plates.

Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving

  • Contains everything needed to prepare low-salt LB medium or agar plates, saving hours of medium preparation time
  • No weighing or mixing of media components
  • No autoclaving
  • No waiting for media to cool before adding antibiotics

Bioproduction, Cloning, Transformation

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Kanamycin
Content And Storage Each type of imMedia™ growth medium is provided in a 20-pouch box. Each pouch contains sufficient reagents to prepare 8-10 standard 100 mm2 agar plates.

Store at room temperature.

All reagents are guaranteed stable for 12 months when properly stored.

Form Powder
Media Type LB, Quick-Preparation Media (e.g. Immedia™)
Preparation Method Microwave
Product Type Bacterial Growth Medium
Target Organism Class E. coli
Final Product Type Agar Plates
Includes Kanamycin
Material Powder Reagents
Packaging Type Ready-to-Use Individual Pouches
Product Line imMedia
Quantity 20 Ea.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.