missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ 2000 CD Transfection Reagent

Product Code. 10194622
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10194622 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10194622 Supplier Invitrogen™ Supplier No. 12566014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Proprietary animal origin-free, chemically defined formulation for transfecting nucleic acids into eukaryotic cells

These reagents allow highly efficient transfection of broad range of cell types, including adherent, suspension, insect, and endothelial lines, as well as primary cultures. All transfection reagents include streamlined protocols for maximum convenience and ease of use in many applications, including high-throughput applications.

 

  • Highest transfection efficiency in many cell types and formats
  • Animal origin-free formulation ensures that mammalian cell culture and bioproduction processes are free of animal-derived materials
  • DNA-Lipofectamine 2000 CD complexes can be added directly to cells in culture medium
  • It is not necessary to remove complexes or change/add medium after transfection, but complexes may be removed after 4–6 hours

 

PCR and Real-Time PCR, Reverse Transcription, cDNA Synthesis Direct from Cells

Order Info

Shipping Condition: Qualified for Ambient or Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free, Chemically Defined
Content And Storage Each reagent should be stored at 2–8°C upon receipt.
Cell Type Established Cell Lines
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 1 mL
Serum Compatible Yes
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.