Learn More
Invitrogen™ Lipofectamine™ LTX Reagent
Description
Includes
Each unit contains one vial of Lipofectamine LTX Reagent (15mL).
Lipofectamine LTX Reagent is an advanced solution for gene expression in both common and challenging cell types. No other plasmid DNA-specific transfection reagent can match the efficiency, convenience, and gentleness of Lipofectamine LTX Reagent for transfection of primary, challenging, and sensitive cell lines. Lipofectamine LTX Reagent is synthesized from 100% animal origin-free components, making it easy to validate the absence of zoonotic diseases, such as BSE or viruses, in research experiments or cell lines.
- Transfect a broad range of cell types, including primary challenging and sensitive cells, with high efficiency
- Combine high transfection efficiency with maximal (>90%) cell viability
- Optimized protocols – rapid, simple protocols provided for most cell lines
- Effective reagent to generate stable cell lines
- Transfect primary cells efficiently
- Superior delivery into challenging cells
- Broad compatibility with cell lines
- Stable transfection
- Simple, rapid, serum-compatible protocol
Cell Culture, Mammalian Cell Culture
Informations sur la commande
Shipping Condition: Wet Ice
Spécification
Spécification
| Classification | Animal Origin-Free |
| Content And Storage | Each unit contains one vial of Lipofectamine™ LTX Reagent (15 mL). Store at 2–8°C. |
| Cell Type | Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
| Sample Type | Plasmid DNA, RNAi Plasmids (shRNA, miR) |
| Transfection Technique | Lipid-based Transfection |
| For Use With (Application) | Transfection |
| High-throughput Compatibility | High-throughput Compatible |
| Product Line | Lipofectamine |
| Product Type | LTX Reagent |
| Afficher plus |
For Research Use Only. Not for use in diagnostic procedures.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.