Learn More
Invitrogen™ Low DNA Mass Ladder
Description
Includes
Low DNAmass™ Ladder, 200μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA
Invitrogen Low DNA Mass Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. Low DNA Mass Ladder consist of six individual chromatography-purified DNA fragments.
Low DNA Mass Ladder is ideal for separation on 1–2% agarose gels.
Highlights of Low DNA Mass Ladder
• Sharp, clear bands—chromatography purified fragments for consistent and reliable results
• Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
• Precise—an exact amount of DNA in each band
Product Use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with DNA bands of increasing intensity. An exact amount of DNA in each band allows approximate quantification of DNA samples.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | • 200 µL Low DNA Mass Ladder • 1 mL10X BlueJuice Gel Loading Buffer Store at -20°C. |
| Concentration | 0.1175 μg/μL |
| Gel Type | Agarose |
| Ready to Load | No |
| Sample Loading Volume | 1 mL |
| Size Range | 100 to 2000 bp |
| Volume (Metric) | 200 μL |
| Gel Compatibility | Agarose gel |
| No. of Reactions | 50 Applications |
| Quantity | 23.5 μg |
| Show More |
Frequently Asked Questions (FAQs)
Sequences of Invitrogen DNA and RNA ladders are proprietary.
Invitrogen DNA ladders contain linear dsDNA fragments.
Invitrogen DNA ladders are composed of double-stranded DNA fragments only.
Here are a few reasons why you might see smearing of the bands:
- The DNA was degraded. Avoid nuclease contamination of DNA standards.
- Too much DNA was loaded on the gel. Decrease the amount of DNA in the gel.
- The DNA was contaminated by protein. Remove proteins by phenol extraction before electrophoresis.
- For small DNA, the bands may have diffused during staining. Add the ethidium bromide before electrophoresis.
- For radiolabeled DNA, labeling was performed by nick translation. Label the DNA by replacement synthesis with T4 DNA polymerase or label the 5' end with T4 polynucleotide kinase.
- Improper electrophoresis conditions were used. Do not allow voltage to exceed ~20 V/cm. Maintain a temperature <30°C during electrophoresis. Check that the electrophoresis buffer used has sufficient buffering capacity.
- The DNA contained too much salt. Remove excess salt by ethanol precipitation before electrophoresis.
This can happen if the marker was heated. Please ensure that the ladders are not heated before use.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.