missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ M13 Forward (-20) Primer

Product Code. 11589146
Change view
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11589146 2 μg 2µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11589146 Supplier Invitrogen™ Supplier No. N52002
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oligonucleotides complementary to a DNA template are necessary to prime DNA synthesis for sequencing reactions.

Oligonucleotides complementary to a DNA template are necessary to prime DNA synthesis for sequencing reactions. We offer a wide selection of Invitrogen single-strand primers that may be used in either single- or double-stranded sequencing protocols. All sequencing primers are non-phosphorylated and are supplied lyophilized.

M13 Forward (-20) Primer features include:

  • Desalted and purified by gel filtration
  • Assayed for function in automated sequencing reactions
  • Supplied in quantity of 2 μg

Order Info

Shipping Conditions: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primer
Content And Storage • M13 Forward (-20) Primer (2 μg)

Store in Freezer at –20°C.
Guaranteed stable for 6 months when properly stored.
Primer Length 16-mer
Primer Sequence 5 ́d[GTAAAACGACGGCCAG]3 ́
Purification Method Desalted, Gel Filtration Purified
Shipping Condition Room Temperature
Primer M13
Quantity 2 μg
For Use With (Application) Sequencing Reactions
Form Lyophilized

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.