missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mimic™ Sf9 Insect Cells

Product Code. 10062692
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10062692 1 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10062692 Supplier Gibco™ Supplier No. 12552014
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Modified to stably express a variety of mammalian glycosyltransferases

Mimic Sf9 Insect Cells are a derivative of the Sf9 insect cell line. Typically, insect cells are unable to process N-glycans to the extent that mammalian cells do. This can affect protein structure, function, antigenicity, and enzymatic activity. The addition of mammalian glycosyltransferases to the Mimic™ Sf9 Insect Cells allows for production of biantennary, terminally sialyated N-glycans from insect cells. The cells can be used to produce more mammalian-like proteins in both baculovirus and stable insect expression systems.

1mL of Mimic Sf9 Insect Cells are provided frozen in 60% complete TNM-FH, 30% FBS, 10% DMSO at a concentration of 107 cells/mL.

Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen.
Cell Line Sf9
Cell Type Insect Cells
Species S. frugiperda
Product Line Mimic
Product Type Insect Cells
Quantity 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.