missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Taq DNA Polymerase, native
A thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer
76.00 € - 2435.00 €
Specifications
Detection Method | Primer-probe |
---|---|
Format | Stand-alone enzyme |
GC-Rich PCR Performance | Low |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Includes: Contains: 20μL Taq DNA Polymerase (5U/μL); 1.25mL 10X PCR buffer (without magnesium); 1mL Magnesium Chloride (50mM)
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10388482
|
Invitrogen™
18038018 |
100 Units |
76.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174512
|
Invitrogen™
18038042 |
500 Units |
269.00 €
500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11538906
|
Invitrogen™
18038067 |
3 x 500 Units |
742.00 €
Pack of 3 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10093102
|
Invitrogen™
18038240 |
5000 Units |
2435.00 €
Each |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The enzyme consists of single polypeptide with molecular weight of 94 kDa. It has 5'-3' DNA polymerase activity and 5'-3' exonuclease activity.
- Amplification of PCR products up to 5 kb in size
- Enzyme that is licensed and qualified for PCR
- Applications: Amplification of DNA from complex genomic, viral, and plasmid templates, RT-PCR, sequencing ssDNA, and cycle sequencing
- Source: Native enzyme is purified from Thermus aquaticus YT1
- Unit definition: One unit of Taq DNA Polymerase is amount of enzyme required to incorporate 10nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C
ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Methylation Analysis, PCR, PCR and Real-Time PCR, RNAi, Epigenetics and Non-Coding RNA Research, Routine PCR
Order Info
Shipping Condition: Approved for shipment on wet or dry ice
Specifications
Primer-probe | |
Low | |
Standard | |
DNA Polymerase | |
1 X | |
3'-A | |
5 kb or less |
Stand-alone enzyme | |
Taq DNA Polymerase | |
5' - 3' | |
5 U/μL | |
No | |
Separate Components | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.