missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Taq DNA Polymerase PCR Buffer (10X)

Product Code. 10043092
Change view
Click to view available options
Quantity:
2 x 1 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10043092 2 x 1 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10043092 Supplier Invitrogen™ Supplier No. 18067017
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Taq DNA Polymerase PCR Buffer is a 10X buffer [200 mM Tris HCl (pH 8.4), 500 mM KCl] supplied with 1 mL of 50 mM MgCl2.

Taq DNA Polymerase PCR Buffer is a 10X buffer supplied with 1 mL of 50 mM MgCl2. It is included with Platinum Taq, Taq, and the SuperScript First-Strand Synthesis System for RT-PCR. This buffer includes 200 mM Tris HCl (pH 8.4) and 500 mM KCl. It is available here to supplement kits where additional buffer may be needed based on protocol variations.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DNA Polymerase PCR Buffer
Concentration 10X
For Use With (Application) Standard PCR
Quantity 2 x 1 mL
Includes • 10X Taq DNA Polymerase PCR Buffer, 2 x 1 mL
• 50 mM MgCl2, 1 mL

Store at -20°C.
Guaranteed stable for 6 months when properly stored.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.