Learn More
Invitrogen™ Neon™ Transfection System
Description
Includes
Neon electroporation device, pipette, pipette station, USB thumbdrive, manual and power cord.
Enables fast and efficient delivery of nucleic acids into all mammalian cell types, including primary, stem, and difficult-to-transfect cells. The flexible and open system allows the user to perform high-quality transfections using optimized or user-defined protocols in three simple steps with as few as 2 × 104 cells per reaction and a reaction time of just 1 second. A novel reaction chamber provides a dramatic increase in transfection efficiency and cell viability.
- Up to 90% in many cell types, including difficult-to-transfect cells, primary, and stem cells
- Easily transfect from 2 × 104 cells to 6 × 106 cells per reaction
- Easy to use, with a single reagent kit for all cell types
- Open system allows electroporation parameters to be optimized freely
- One-of-a-kind electroporation
- Plug-n-play out of the box
- Perform more transfections with limited or precious cells
For use at room temperature under normal laboratory conditions.
Adenoviral shRNA, Cell Culture, Lentiviral shRNA & miRNA, Plasmid Transfection, RNAi, RNAi Transfection, RNAi, Epigenetics & Noncoding RNA Research, Retroviral RNAi, Stem Cell & Primary Cell Transfections, Synthetic siRNA Transfection, Transfection, Vector-Based RNAi, siRNA
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Amperage | 3.0-6.0 A |
| Classification | Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum |
| Content And Storage | The Neon™ Transfection System comes with a Neon™ electroporation device, pipette, pipette station, USB thumbdrive, manual and power cord. For use at room temperature under normal laboratory conditions. |
| Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
| Sample Type | Plasmid DNA, Synthetic siRNA |
| For Use With (Equipment) | Neon™ Transfection System |
| For Use With (Application) | Transfection |
| Max. Voltage | 0.5-2.5 kV |
| Product Line | Neon |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.