Learn More
Invitrogen™ pAd/BLOCK-iT™-DEST RNAi Gateway Vector
Beschreibung
Benötigt
Additional materials required, available separately: Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Invitrogen offers state-of-the-art Gateway destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins. All Gateway destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an Ultimate™ RF Clone. A wide range of destination vectors is available.
- All destination vectors are provided lyophilized and supercoiled
Adenoviral shRNA, Cloning, Gateway Cloning, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Virus-Based RNAi
Bestellinformation
Shipping Condition: Dry Ice
Spezifikation
Spezifikation
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Adenoviral |
| Promoter | U6 |
| Product Type | RNAi Gateway Vector |
| Content And Storage | All destination vectors are provided lyophilized and supercoiled. |
| Cloning Method | Gateway |
| Quantity | 6 μg |
| Vector | pAd, pDEST |
| Product Line | BLOCK-iT, Gateway |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.