missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pcDNA™4/HisMax A, B, & C Mammalian Expression Vectors

Product Code. 10553423
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10553423 20 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553423 Supplier Invitrogen™ Supplier No. V86420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

The pcDNA4/HisMax A, B and C vectors, 20μg each, are supercoiled and provided lyophilized. 20μg of pcDNA4/HisMax/lacZ control plasmid is also provided. The pcDNA4/HisMax TOPO TA Expression Kit contains two boxes. The TOPO TA box contains 200ng of linearized and topoisomerase I-activated pcDNA4/HisMax-TOPO vector, dNTPs, control PCR template and primers, forward and reverse primers for sequencing and PCR screening, and a lacZ expression control plasmid. The box contains transformation reagents including 21 x 50μL aliquots of TOP10 E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid.

Designed to maximize protein expression in a variety of mammalian cells

  • Contains QBI SP163 translational enhancer to increase expression levels two- to five-fold above those seen with promoter alone
  • In addition to SP163-enhanced expression, pcDNA4/HisMax includes cleavable N-terminal Xpress™ tag for rapid detection of recombinant protein with Anti-Xpress Antibody

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Transfection
Promoter CMV
Product Type Mammalian Expression Vector
Selection Agent (Eukaryotic) Zeocin™
Content And Storage

The pcDNA™4/HisMax A, B, & C vectors, 20 μg each, are supercoiled and provided lyophilized. 20 μg of pcDNA™4/HisMax/lacZ control plasmid is also provided. Store vectors at -20°C. The pcDNA™4/HisMax TOPO™TA Expression Kit contains two boxes. The TOPO™ TA box contains 200 ng of linearized and topoisomerase I-activated pcDNA4/HisMax-TOPO™ vector, dNTPs, control PCR template and primers, forward and reverse primers for sequencing and PCR screening, and a lacZ expression control plasmid.
Store this box at -20°C. The One Shot™ box contains transformation reagents including twenty-one 50-μl aliquots of TOP10 E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid. The One Shot™ box should be stored at -80°C. Guaranteed stable for 6 months when properly stored.

Cleavage EK (Enterokinase) Recognition Site
Protein Tag His Tag (6x), Xpress Epitope Tag
Cloning Method Restriction Enzyme/MCS
Quantity 20 μg
Vector pcDNA
Product Line pcDNA
For Use With (Application) Constitutive Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.