missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pcDNA™5/FRT Mammalian Expression Vector
Description
Includes
Both pcDNA5/FRT and pcDNA5/FRT/CAT vectors.
- This vector features CMV promoter for high level expression in mammalian cells
- Ten unique restriction sites for easy cloning
- FLP Recombination Target (FRT) site for Flp recombinase-mediated integration of vector into Flp-In host cell line
- Hygromycin resistance gene for selection of stable cell lines
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Mammalian Expression Vector |
| Selection Agent (Eukaryotic) | Hygromycin |
| Content And Storage | Contains both pcDNA5⁄FRT and pcDNA5⁄FRT⁄CAT vectors. Store at -20°C |
| Protein Tag | Untagged |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 20 μg |
| Vector | pcDNA |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?