missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pEF6/V5-His TOPO™ TA Expression Kit

Product Code. 10482662
Change view
Click to view available options
Quantity:
20 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10482662 20 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482662 Supplier Invitrogen™ Supplier No. K961020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Each pEF6/V5-His TOPO TA Expression Kit contains two boxes. The TOPO TA box contains all of the reagents necessary for ligation, including 200ng of topoisomerase I-activated pEF6/V5-His-TOPO vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control templates and primers, primers for sequencing or PCR screening, and an expression control plasmid. The Competent Cell box contains all of the reagents necessary for transformation, including 21 x 50μL aliquots of TOP10 Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid.

Designed for high-level mammalian expression from the powerful human EF-1 α promoter with the added convenience of one-step TOPO Cloning

  • Enhancer/promoter elements from human elongation factor 1α subunit (hEF-1α) for high-level expression in mammalian cells
  • Vector includes Blasticidin resistance gene for rapid selection of stable mammalian cell lines
  • A C-terminal V5 epitope for efficient detection with an Anti-V5 Antibody
  • A C-terminal polyhistidine (6xHis) sequence for rapid purification with nickel-chelating resin and detection with Anti-His(C-term) Antibody

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter EF-1α
Product Type TOPO TA Expression Kit
Selection Agent (Eukaryotic) Blasticidin
Content And Storage

Each pEF6/V5-His TOPO™ TA Expression Kit contains two boxes. The TOPO™ TA box contains all of the reagents necessary for ligation, including 200 ng of topoisomerase I-activated pEF6/V5-His-TOPO™ vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control templates and primers, primers for sequencing or PCR screening, and an expression control plasmid. Store at -20°C. The One Shot™ Competent Cell box contains all of the reagents necessary for transformation, including twenty-one 50-μl aliquots of TOP10 Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid. Store at -80°C. Guaranteed stable for 6 months when properly stored.

Cleavage EK (Enterokinase) Recognition Site
Protein Tag His Tag (6x), V5 Epitope Tag
Cloning Method TOPO-TA
Quantity 20 reactions
Vector pEF, TOPO-TA Vectors
Product Line TOPO
For Use With (Application) Constitutive Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.