Learn More
Invitrogen™ pGlow TOPO™ TA Expression Kit
Description
Includes
pGlow TOPO TA Expression Kits contains two boxes. The TOPO TA Cloning box contains linearized and topoisomerase I-activated pGlow-TOPO vector, dNTPs, primers for sequencing, salt solution, and control PCR template and primers. The box contains TOP10 E. coli cells, S.O.C. medium, and a supercoiled control plasmid.
- Cycle 3 GFP is exellent for noninvasive analysis of promoter elements within intact, living cells Fluorescent property of Cycle 3 GFP allows
- in vivo detection in virtually any cell type or species using microscopy with wild-type GFP filter sets or by fluorescence-activated cell sorting methods
Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP)/Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays
Specifications
Specifications
| Delivery Type | Transfection |
| Promoter | None (Promoterless) |
| Product Type | TOPO TA Expression Kit |
| Reporter Gene | GFP (Cycle 3) |
| Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
| Content And Storage | pGlow TOPO™ TA Expression Kits contains two boxes. The TOPO™ TA Cloning box contains linearized and topoisomerase I-activated pGlow-TOPO™ vector, dNTPs, primers for sequencing, salt solution, and control PCR template and primers; store at -20°C. The One Shot™ box contains One Shot™ TOP10 E. coli cells, S.O.C. medium, and a supercoiled control plasmid; store at -80°C. The kits are guaranteed stable for 6 months when properly stored. |
| Protein Tag | Untagged |
| Cloning Method | TOPO-TA |
| Quantity | 20 reactions |
| Vector | pGlow, TOPO-TA Vectors |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.