missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase

Product Code. 10277202
Change view
Click to view available options
No. of Reactions:
120 Reactions
300 Reactions
5000 Reactions
600 Reactions
Unit Size:
120 reactions
300 reactions
5000 reactions
600 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10277202 300 Reactions 300 reactions
10033232 120 Reactions 120 reactions
10358742 600 Reactions 600 reactions
10728473 5000 Reactions 5000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10277202 Supplier Invitrogen™ Supplier No. 10966026
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Each box contains: Platinum Taq DNA Polymerase; 10X PCR Buffer (no magnesium chloride); 50mM magnesium chloride; KB Extender

Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable 'hot start' thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase.

Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable hot-start thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The hot start property of the enzyme is conferred by thermolabile monoclonal antibodies that render Taq DNA polymerase inactive until the initial PCR denaturation step, thus preventing the extention of nonspecifically annealed primers and improving product yield.

The hot-start property allows for convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity. PCR products generated with Platinum Taq DNA Polymerase may be used in the same downstream applications without protocol modifications.

Features

  • Convenient reaction assembly at room temperature
  • 5' to 3' exonuclease activity
  • Lacks 3' to 5' exonuclease activity

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • Hot-start PCR
  • RT-PCR

Notes

  • For superior PCR performance, the next-generation enzyme Platinum II Taq Hot-start DNA Polymerase is recommended.
  • Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.

Order Info

Shipping Condition: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Platinum Taq DNA Polymerase, 60 μL
• 10X PCR Buffer (without magnesium), 2 x 1.25 mL
• 50 mM MgCl2, 1 mL
• KB Extender, 1.25 mL

Store at -10°C to -30°C in a non-frost-free freezer.
Format Tube
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Quantity 300 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 300 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.