Learn More
Invitrogen™ Platinum™ II Hot-Start PCR Master Mix (2X)
Invitrogen™ Hot-Start PCR Master Mix for fast and easy PCR
85.75 € - 1532.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15889844
|
Invitrogen™
14000012 |
50 Reactions |
85.75 €
50 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899844
|
Invitrogen™
14000013 |
200 Reactions |
334.00 €
200 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15809854
|
Invitrogen™
14000014 |
1000 Reactions |
1532.00 €
1000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Invitrogen™ Platinum™ Taq II Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq, and leading hot-start technology.
- Innovative buffer—enables universal primer annealing protocol by isostabilizing primer-template duplex structures
- Engineered Taq—confers fast cycling and tolerance to common inhibitors
- Platinum hot-start technology—ensures superior specificity, sensitivity, and yields; allows for room temperature reaction setup
- Optional green buffer—reduces pipetting error with direct gel loading
Additional:
- Antibody-mediated hot-start
- Universal annealing protocool
- Inhibitor resistance
Applications:
- Genotyping
- High-throughput PCR
- GC-rich templates
- Fast PCR
- Amplification of samples with suboptimal purity
- Direct PCR with blood
- PCR from DNA extracted from FFPE samples
- Colony PCR
- PCR from bacterial 16S rRNA genes
Specifications
Tube | |
High |
For Research Use Only. Not for use in diagnostic procedures.