missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG w/ROX

Product Code. 11538656
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11538656 500 Reactions 500 reactions
11528656 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11538656 Supplier Invitrogen™ Supplier No. 11743500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Platinum™ Quantitative PCR SuperMix-UDG with ROX; Magnesium Chloride (50mM)

qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.

qPCR SuperMixes with ROX are available in two formats:
  • Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye
  • Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)

Order Info

Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 12.5 mL Platinum™ Quantitative PCR SuperMix-UDG with ROX
• 2 × 1 mL Magnesium Chloride (50 mM)

Store at -20°C in the dark.
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System
Product Line Platinum
Product Type Quantitative PCR SuperMix-UDG
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions
For Use With (Application) Gene Expression
Fidelity (vs. Taq) 2 X
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Volume 12.5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.