missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellsDirect™ One-Step qRT-PCR Kit with ROX, for ABI 7000, 7300, 7700, and 7900(HT) instruments

Product Code. 10583513
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10583513 500 Reactions 500 reactions
10504863 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10583513 Supplier Invitrogen™ Supplier No. 11754500
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The CellsDirect One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

The CellsDirect One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.

Streamline Your Workflow
In traditional real-time quantitative RT-PCR (qRT-PCR), you first isolate RNA from cells in a time-consuming procedure that can lead to loss of material. Using the CellsDirect One-Step qRT-PCR Kit, you lyse the cells and add the complete lysate directly to a one-step qRT-PCR reaction with minimal handling and sample loss.

Ideal for Your qRT-PCR Application
CellsDirect technology is validated for gene expression studies involving limited samples, and is suitable for validation of RNAi knockdowns. CellsDirect One-Step qRT-PCR Kits are especially valuable for high-throughput applications by eliminating costly, time-consuming RNA purification steps. The CellsDirect One-Step qRT-PCR Kit may also be used to detect genomic DNA targets.

CellsDirect One-Step qRT-PCR Kits may be used in conjunction with hydrolysis probes (e.g., TaqMan™) or LUX™ fluorogenic primers.

  • Efficiency: Offers significant time and cost savings for increased throughput
  • Convenience: Enzyme mix includes both SuperScript™ III Reverse Transcriptase and Platinum™ Taq DNA Polymerase
  • Sensitivity: No separate RNA isolation step increases sensitivity by minimizing sample loss
  • Broad Detection Range: Detect targets from samples ranging from a single cell to 10,000 cells
  • Versatility: Compatible with a wide variety of cell lines and samples collected by Laser Capture Microdissection (LCM)
  • For use with ABI 7000, 7300, 7700, and 7900(HT) instruments

This kit includes 2X Reaction Mix containing ROX™ Reference Dye. An equivalent kit containing a separate tube of ROX Reference Dye, is also available.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Resuspension Buffer, 10 mL
• Lysis Enhancer, 1 mL
• 1 U/μL DNase I, Amplification Grade, 2 x 1.25 mL
• 10X DNase I Buffer, 800 μL
• 25 mM EDTA, 2 x 1 mL
• SuperScript™ III RT/Platinum™ Taq Mix, 500 μL
• 2X Reaction Mix with ROX™, 12.5 mL
• 50 mM MgSO4, 1 mL
• DEPC-treated water, 12.5 mL
• 10 ng/μL HeLa Total RNA, 10 μL

The kit is shipped on dry ice and should be stored at -20°C.
Detection Method Primer-probe
Reaction Speed Fast or Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, QuantStudio 3 and 5 systems, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Product Line CellsDirect, Platinum, SuperScript
Quantity 500 reactions
For Use With (Application) RT-PCR
No. of Reactions 500 Reactions
Sample Type Cells

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.