missing translation for 'onlineSavingsMsg'
Learn More

Platinum™, SYBR™ Platinum™ SYBR™ Green qPCR SuperMix-UDG

Product Code. 10633863
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Conditionnement:
500 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit No. of Reactions Conditionnement
10633863 500 Reactions 500 reactions
10623863 100 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 10633863 Fournisseur Platinum™, SYBR™ Code fournisseur 11733046
 Restricted Item
In Stock
Ajouter au panier
Edge
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Includes

Platinum SYBR Green qPCR SuperMix-UDG (2 × 1.25mL), 50mM Magnesium Chloride (1mL), ROX Reference Dye (100μL), 20X BSA (300μL)

Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.

The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.

  • Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
  • 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
  • Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
  • SYBR™ Green I dye for simple and easy detection of real-time product

Informations sur la commande

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Spécification

Content And Storage • Platinum™ SYBR™ Green qPCR SuperMix-UDG (12.5 mL)
• 50 mM Magnesium Chloride (2 × 1 mL)
• ROX Reference Dye (500 μL)
• 20X BSA (1.3 mL)

Sufficient reagents are provided for 500 reactions of 50 μL each. Stored at -20°C.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ
Product Line Platinum, SYBR
Product Type Real Time PCR SYBR Master Mix
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions of 50 μL Each
For Use With (Application) Gene Expression
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Volume 12.5 mL
Afficher plus Afficher moins

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.