missing translation for 'onlineSavingsMsg'
Learn More

SuperScript™, Platinum™, SYBR™ SuperScript™ III Platinum™ SYBR™ Green One-Step qPCR Kit w/ROX

Product Code. 11588656
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11588656 100 Reactions 100 reactions
11598656 500 Reactions 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11588656 Supplier SuperScript™, Platinum™, SYBR™ Supplier No. 11746100
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Combine a powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.

One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. SuperScript™ III Reverse Transcriptase produces higher yields of full-length cDNA for broader gene representation. Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. ROX reference dye premixed at optimal concentration for ABI instruments.

  • Provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store in freezer (-5 to -30°C).
Detection Method SYBR
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System
Product Line Platinum, SYBR, SuperScript
Product Type Real Time PCR SYBR Master Mix
Quantity 100 reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
No. of Reactions 100 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.