missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum™, ThermoScript™ Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System
Description
Includes
ThermoScript RT/Platinum Taq mix, and 2X ThermoScript reaction mix (buffer containing 0.4mM of each dNTP, 6mM MgSO4 ), vial of 50mM MgSO4
This kit provides Platinum™ Taq DNA Polymerase, ThermoScript RT polymerase and a vial of 50mM MgSO4
Order Info
Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice
Specifications
Specifications
| Content And Storage | Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System contains ThermoScript™ RT/Platinum™ Taq mix, and 2X ThermoScript™ reaction mix (a buffer containing 0.4 mM of each dNTP, 6 mM MgSO4 ). A vial of 50 mM MgSO4 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored. |
| Detection Method | Primer-probe |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| Product Line | Platinum, ThermoScript |
| Product Type | RT-PCR One-Step System |
| Quantity | 500 reactions |
| Shipping Condition | Dry Ice |
| Sufficient For | 500 Reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?