missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti6/TR Vector Kit

Product Code. 10297092
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10297092 1 Kit 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10297092 Supplier Invitrogen™ Supplier No. V48020
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

The pLenti6/TR Vector Kit includes 20μg of pLenti6/TR vector and 50mg of Blasticidin.

Make stable cell lines expressing high levels of tetracycline repressor (TR) protein under the control of the CMV promoter

This vector contains elements that allow packaging of the construct into lentiviral particles and the Blasticidin resistance marker for selection of stable cell lines. The pLenti6/TR vector is designed for use with the BLOCK-iT™ Inducible H1 Lentiviral RNAi System, BLOCK-iT Inducible H1 RNAi Entry Vector Kit, and the ViraPower™ T-REx™ Lentiviral Expression System.

Lentiviral shRNA and miRNA, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Regulated Expression, Viral Gene Delivery, Virus-Based RNAi

TRUSTED_SUSTAINABILITY

Specifications

Product Type RNAi Expression Vector Kit
Content And Storage The pLenti6/TR Vector Kit includes 20 μg of pLenti6/TR vector and 50 mg of Blasticidin. Store at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Cloning Method Gateway™
Constitutive or Inducible System Inducible
Delivery Type Lentiviral
Selection Agent (Eukaryotic) Blasticidin
Vector pLenti
Promoter CMV
Format Kit
Product Line BLOCK-iT
For Use With (Application) Viral Expression
Quantity 1 Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.