missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti6/V5 Directional TOPO™ Cloning Kit

Product Code. 10562693
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10562693 20 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10562693 Supplier Invitrogen™ Supplier No. K495510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Directional TOPO Cloning site for rapid and efficient directional cloning of blunt-end PCR products

  • Contains TOPO-adapted ViraPower lentiviral expression vector, pLenti6/V5-D-TOPO for quick PCR-based cloning and expression of a target gene in dividing and nondividing mammalian cells
  • Vector has CMV promoter for driving high-level, constitutive expression of the target gene and blasticidin selection marker for stable selection in mammalian cells
  • High efficiency and rapid cloning
  • Produces replication-incompetent virus for enhanced biosafety of system
  • Rouse Sarcoma Virus (RSV) enhancer/promoter for Tat-independent production of viral mRNA in the producer cell line
  • Modified HIV-1 5' and 3' Long Terminal Repeats (LTR) for viral packaging and reverse transcription
  • HIV-1 psi (ψ) packaging sequence for viral packaging
  • HIV Rev response element (RRE) for Rev-dependent nuclear export of unspliced viral mRNA
  • (CMV) immediate early promoter for high-level constitutive expression of gene of interest in mammalian cells
  • C-terminal V5 epitope for detection of recombinant protein
  • Blasticidin (bsd) resistance gene for selection in E. coli and mammalian cells
  • Ampicillin resistance gene for selection in E. coli
  • pUC origin for high-copy replication and maintenance of plasmid in E. coli

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery

TRUSTED_SUSTAINABILITY

Specifications

Cloning Method Directional TOPO™
Constitutive or Inducible System Constitutive
Content And Storage pLenti⁄V5-D-TOPO™ reagents (Box 1) Store at -20°C
• pLenti6⁄V5-D-TOPO™ Reagents (15-20 ng⁄μl linearized plasmid DNA in: 50% glycerol, 50 mM Tris-HCl, pH 7.4 (at 25°C),1 mM EDTA, 2 mM DTT, 0.1% Triton X-100, 100 μg⁄ml BSA, 30 μM bromophenol blue)
• dNTP Mix (12.5 mM each dATP, dCTP, dGTP, dTTP in water, pH8 at a total volume of 10 μl)
• Salt Solution (1.2 M NaCl and 0.06 M MgCl2, volume of 50 μl)
• Water (1 ml)
• CMV Forward Sequencing Primer (0.1 μg⁄μl in TE Buffer, pH 8, pH8 at a total volume of 20 μl)
• Control PCR Primers (0.1 μg⁄μl each in TE Buffer, pH 8, pH8 at a total volume of 10 μl)
• Control PCR Template (0.1 μg⁄μl in TE Buffer, pH 8, pH8 at a total volume of 10 μl)
• pLenti6⁄V5-GW⁄lacZ (lyophilized in TE Buffer, pH 8, 10 μg)

One Shot™ Stbl3™ Chemically Competent E. coli (Box 2) Store at -80°C
• S.O.C. Medium (2% Trypton, 0.5% Yeast Extract, 10 mM NaCl, 2.5 mM KCl, 10 mM MgCl2, 10 mM MgSO4, 20 mM glucose. 6ml) May be stored at +4°C or room temperature
• Stbl3™ Cells (21 x 50 μl)
• pUC19 Control DNA (10 pg⁄μl in 5 mM Tris-HCl, 0.5 mM EDTA, pH 8. 50 μl)
Delivery Type Lentiviral
For Use With (Application) Viral Expression
No. of Reactions 20 reactions
Product Line TOPO, ViraPower
Product Type TOPO Cloning Kit
Promoter CMV
Protein Tag V5 Epitope Tag
Quantity 20 Reactions
Selection Agent (Eukaryotic) Blasticidin
Vector pLenti, Directional TOPO Vectors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.