missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLUS™ Reagent

Product Code. 10287052
Change view
Click to view available options
Quantity:
0.85 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10287052 0.85 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10287052 Supplier Invitrogen™ Supplier No. 11514015

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

Contains one vial (0.85mL) PLUS Reagent.

Used in conjunction with various transfection reagents, such as Lipofectamine™ Reagent, to improve transfection efficiency in adherent cell lines

  • Some customers find that when using PLUS Reagent, less optimization of DNA concentration is needed to achieve high transfection performance
  • Addition of PLUS Reagent may also reduce quantities of DNA and lipid required by approximately one-half compared to lipid alone
  • PLUS Reagent was specifically designed to expand array of transfectable cell types
  • When used with Lipofectamine Reagent, PLUS Reagent has been demonstrated to improve the transfection of BHK-21, NIH 3T3, and other cell lines

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one vial (0.85 ml) PLUS™ Reagent. Store at 4°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, RNAi Plasmids (shRNA, miR)
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line PLUS
Product Type Reagent
Quantity 0.85 mL
Serum Compatible Yes
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.