missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K, recombinant

Product Code. 10124532
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10124532 1 g 1g
11578916 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124532 Supplier Invitrogen™ Supplier No. 25530031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range–optimal activity between 6.5 and 9.5
  • Under denaturing conditions–e.g., in the presence of SDS or urea
  • In the presence of metal chelating agents–e.g., EDTA
  • At comparatively high temperatures–optimum digestion temperature is 65°C
  • Protein Filter: P

Applications

Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing

Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition

One mAnson unit is described as that amount of enzyme that liberates 1μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥40 U/mg
Content And Storage One tube Proteinase K
Store in refrigerator (2–8°C).
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Quantity 1 g
Product Type Proteinase K
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.