missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K, recombinant

Product Code. 11578916
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11578916 100 mg 100mg
10124532 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11578916 Supplier Invitrogen™ Supplier No. 25530015
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range–optimal activity between 6.5 and 9.5
  • Under denaturing conditions–e.g., in the presence of SDS or urea
  • In the presence of metal chelating agents–e.g., EDTA
  • At comparatively high temperatures–optimum digestion temperature is 65°C
  • Protein Filter: P

Applications

Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing

Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition

One mAnson unit is described as that amount of enzyme that liberates 1μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One vial

Store in refrigerator (2–8°C).
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Quantity 100 mg
Product Type Proteinase K
Form Powder
How do you recommend storing proteinase K?

We recommend storing it at 4°C.

How do I isolate DNA from single bacterial colonies before PCR?

Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.