missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pSCREEN-iT™/lacZ-DEST Gateway™ Vector Kit

Product Code. 10318492
Change view
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10318492 20 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10318492 Supplier Invitrogen™ Supplier No. V47020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For RNAi target screening

Use the pSCREEN-iT™/lacZ-DEST Gateway™ Vector Kit to assess the potency of any RNAi knockdown reagent using a fast, sensitive, and cost-effective β-gal reporter assay or identify the best knockdown reagent for RNAi experiments without knowing protein function, western blotting, or qRT-PCR.
How it Works: The BLOCK-iT RNAi Target Screening system method uses a β-galactosidase activity assay to measure the knockdown ability of an RNAi reagent (Stealth RNAi™ siRNA, siRNA, dicer pools, or shRNA-containing plasmid). To identify the most effective knockdown reagent for your target, simply clone your target gene into the pSCREEN-iT /lacZ-DEST Gateway vector. Co-transfect the resulting construct with the knockdown reagent being tested. Expressed genes will be fused to β-galactosidase, enabling you to use β-galactosidase levels to measure the amount of RNAi-induced degradation of the target gene. The more effective any specific knockdown reagent is at mediating RNAi, the more degradation of the fusion mRNA will occur, lowering β-gal activity.
The BLOCK-iT RNAi Target Screening System includes the pSCREEN-iT /lacZ-DEST Gateway vector for generating a construct containing a lacZ/gene of interest fusion. This vector features:

  • attR sites immediately following the lacZ ORF for fast and efficient recombination with any attL-flanked Gateway entry vector to generate the lacZ/target gene fusions
  • The CMV promoter for strong expression of the lacZ/target gene fusion, leading to increased assay sensitivity and better discrimination between knockdown reagents of varied effectiveness
  • Positional cloning to fuse a gene or fragment to the 3' end of the lacZ gene so that β-galactosidase will be expressed even if the target fragment being tested contains stop codons
In addition, the pSCREEN-iT /lacZ-DEST vector is fully compatible with the Ultimate™ ORF Clone Collection. This makes a wide selection of potential target genes readily available for generating lacZ fusions.

RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Vector-Based RNAi

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Promoter CMV
Product Type β-Galactosidase Reporter Gene Assay System
Reporter Gene Beta-Gal (lacZ)
Content And Storage The pSCREEN-iT™ /lacZ-DEST Gateway™ Vector Kit contains 6 μg of vector and 10 μg of pSCREEN-iT™/lacZ-GW/CDK2 control vector, the positive control Stealth™RNAi lacZ, a negative control Scrambled Stealth RNAi™ siRNA, and 1X annealing/dilution buffer. Store at -20°C. The BLOCK-iT™ RNAi Target Screening Kit contains the pSCREEN-iT™/lacZ-DEST Gateway™ Vector Kit, 250 μl of Lipofectamine™ 2000 Reagent, and the FluoReporter™ lacZ/Galactosidase Quantification Kit. Store the Lipofectamine™ 2000 Reagent at +4°C. Store the FluoReporter™ Kit at -20°C protected from light. The BLOCK-iT™ RNAi Target Screening System contains the BLOCK-iT™ RNAi Target Screening Kit, LR Clonase™ enzyme mix, and pCR ¤ 8/GW/TOPO ¤ TA Cloning Kit. Store the LR Clonase™ enzyme mix at -80°C. The pCR™ 8/GW/TOPO™ TA Cloning Kit includes the ready-to-use pCR™ 8/GW/TOPO™ vector, dNTPs, salt solution, water, forward and reverse sequencing primers, control template and PCR primers, One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a pUC19 control plasmid. Store the Competent Cells at -80°C. Store all other components at -20°C. All components are guaranteed stable for 6 months when properly stored.
Sample Type Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Quantity 20 Reactions
Product Line BLOCK-iT, Gateway
For Use With (Application) RNAi Target Screening

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.