missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic Digestion Buffer

Product Code. 10468843
Change view
Click to view available options
Quantity:
70 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10468843 70 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10468843 Supplier Invitrogen™ Supplier No. K182301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

PureLink Genomic Spin Columns, Collection Tubes, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual. The PureLink 96 Genomic DNA Kit includes PureLink Genomic Binding Plates, Wash Plates, Deep Well Plates, Foil Tape, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual.

The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.

The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.

  • Obtain higher gDNA yields and purity—Optimized spin-column membrane and buffer formulations
  • Save time and money—Use with large range of sample sizes (e.g., 100 μL to 1 mL blood); 96-well version does not require special equipment
  • Increase flexibility—one kit works with variety of sample types

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Digestion Buffer
Column Type Spin Column
Content And Storage Store at room temperature.
Format Kit
Sample Type Bacteria, Blood, Blood Spot, Cells, Swab, Tissue
Product Type Genomic Digestion Buffer
Quantity 70 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.