missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic Lysis/Binding Buffer

Product Code. 10206463
Change view
Click to view available options
Quantity:
80 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10206463 80 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206463 Supplier Invitrogen™ Supplier No. K182302

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

PureLink Genomic Spin Columns, Collection Tubes, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual. The PureLink 96 Genomic DNA Kit includes PureLink Genomic Binding Plates, Wash Plates, Deep Well Plates, Foil Tape, Digestion Buffer, Lysis/Binding Buffer, Wash Buffers, Elution Buffer, Proteinase K, RNase A, and the manual.

The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.

The PureLink™ Genomic DNA Mini Kit and PureLink™ 96 Genomic DNA Kit enable high-yield, high-purity DNA extractions from wide variety of sample types, including blood, tissues, cells, bacteria, swabs, and blood spots, in familiar silica spin-column or semi-skirted 96-well plate format.

  • Obtain higher gDNA yields and purity—Optimized spin-column membrane and buffer formulations
  • Save time and money—Use with large range of sample sizes (e.g., 100 μL to 1 mL blood); 96-well version does not require special equipment
  • Increase flexibility—one kit works with variety of sample types

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer, Binding Buffer
Column Type Spin Column
Content And Storage Store at room temperature.
Format Kit
Sample Type Bacteria, Blood, Blood Spot, Cells, Swab, Tissue
Product Line PureLink
Product Type Genomic Lysis/Binding Buffer
Quantity 80 mL
Shipping Condition Room Temperature
compliance-icons
Product Identifier
  • PURELINK GENOMIC LYSIS/BINDING BUFFER 80ml
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Guanidine hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.