missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Plasmid Maxiprep Kit

Product Code. 10257072
Change view
Click to view available options
Quantity:
10 Preps
25 Preps
Unit Size:
10 preps
25 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10257072 10 Preps 10 preps
10464402 25 Preps 25 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10257072 Supplier Invitrogen™ Supplier No. K210006
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PureLink HiPure Plasmid Maxiprep Kit is designed to isolate transfection-grade plasmid DNA from E. coli.

The PureLink HiPure Plasmid Maxiprep Kit is designed to isolate transfection-grade plasmid DNA from E. coli. The Maxiprep Kit protocol will typically yield 500–850 μg plasmid DNA from 100–200 mL bacterial culture, at purity that is comparable to that achieved by two passes through cesium chloride gradients.

  • Labor and time saving- no need for extra steps to remove endotoxin and other contaminants
  • High-purity, low-endotoxin product- pure enough for mammalian cell transfection
  • Versatility—purify any type and size of plasmid DNA, including BAC, bacmids, and ssM13 DNAs

Anion-exchange chromatography for plasmid purification

  • PureLink HiPure Plasmid DNA Purification Kits use patented anion-exchange resin to purify plasmid DNA to a level equivalent to two passes through CsCl gradients
  • Resin combines excellent capacity with a fast flow rate, high resolution, high yield, and efficient endotoxin removal
  • Plasmid preparations can typically be completed in less than 2 hours

Contaminant-free DNA

  • Unlike cesium chloride gradient protocols, PureLink HiPure Plasmid Purification Systems do not use troublesome solvents to work with and to dispose of
  • Typically, plasmid DNA prepared using the PureLink HiPure Plasmid Midiprep Kit will have an A260/A280 ratio of >1.80, indicating that the DNA is reasonably free of proteins that might interfere with downstream applications
  • Endotoxin levels are typically within 0.1–1 EU/μg, making this plasmid DNA ideal for mammalian cell transfection
  • Purification time is two hours

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 100 mL Resuspension Buffer (R3)
• 550 μL RNase A
• 100 mL Lysis Buffer (L7)
• 100 mL Precipitation Buffer (N3)
• 300 mL Equilibration Buffer (EQ1)
• 2 × 300 mL Wash Buffer (W8)
• 200 mL Elution Buffer (E4)
• 30 mL TE Buffer
• 10 HiPure Columns
• 3 Column Holders

Store all components at room temperature.
Format Column
Isolation Technology Anion Exchange Resin
Scale Maxi
Sample Type Bacterial Culture
Elution Volume 15 mL
Final Product Type BAC DNA, Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 10 Preps
Plasmid <40kb, Low Copy Plasmid, High Copy Plasmid, BAC
Prep Scale > 200 μg (Large-Scale) Plasmid DNA
Product Line PureLink
Product Type Plasmid MaxiPrep Kit
Quantity 10 Preps
Purity Transfection Grade
Shipping Condition Room Temperature
Target BAC DNA, Plasmid DNA
System Type PureLink™
Test Time 2 hr.
Yield 1000 μg
Show More Show Less
compliance-icons
Product Identifier
  • PureLink HiPure Plasmid Maxiprep Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: acetic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.