Learn More
Invitrogen™ PureLink™ PCR Micro Kit
Beschreibung
Beinhaltet
Columns, binding and wash buffers, recovery tubes, and the manual
PureLink™ PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.These highly purified DNA fragments and amplicons provide excellent results in downstream applications such as DNA sequencing, restriction digestion, cloning, PCR, and labeling. PureLink™ PCR Micro Kits provide a simple and rapid method for purifying and concentrating DNA fragments ranging in size from 100 bp-12 kb.
The unique micro-nozzle design allows up to 95% recovery of DNA fragments with as little as a 5μL elution volume. With less than a 10% coefficient of variance in the elution volume you can be sure that no ethanol is carried over from the wash steps giving you the best chance of success in downstream applications. Removal of primers, dNTPs, enzymes, and salts exceeds 98.5%. This kit can also efficiently remove unincorporated dye-labeled nucleotides from a cDNA labeling reaction.
Not High Throughput-Compatible (Manual)
Bestellinformation
Shipping Condition: Room Temperature
Spezifikation
Spezifikation
| Content And Storage | Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature. |
| Format | Spin Column |
| Elution Volume Max | 20 μL |
| Elution Volume Min | 5 μL |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Line | PureLink |
| Product Type | PCR Micro Kit |
| Quantity | 50 Preps |
| Run Time | 10 min. |
| Separation Range | 100 bp to 12 kb |
| Mehr anzeigen |
Frequently Asked Questions (FAQs)
The size cut-off is 100 bp to 12 kb.
The size range is between 100 bp and 12 kb. Our kit comes with two buffers, Binding Buffer (B2) and Binding Buffer HC (B3). The Binding Buffer HC (B3) eliminates primer-dimers and short failed PCR products that are smaller than 300 bp.
Sicherheit und Handhabung
- PCR Micro Kit
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Guanidine hydrochloride
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.