missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RadPrime DNA Labeling System

Product Code. 10780194
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10780194 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10780194 Supplier Invitrogen™ Supplier No. 18428011
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for rapid preparation (under 10 min) of high-specific activity 32 P-labeled probes for the detection of DNA and RNA

  • Produces probes that can detect nucleic acids on northern or Southern blots, plaque lifts, colony hybridizations, and in situ hybridization
  • Yields >109cpm/μg control DNA using [32P]-dCTP
  • Labels 25ng of DNA in one reaction
  • Performance and Quality Testing: Incorporation of a radioactively labeled nucleotide is verified using control DNA in a RadPrime labeling reaction

DNA and RNA Purification and Analysis, DNA Labeling, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Nucleic Acid Labeling and Oligo Synthesis, Southern Blotting

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Labeling System
Detection Method Radioactive
Content And Storage Components of the RadPrime DNA Labeling System are listed in the table to the right. Store at -20°C.
.
Final Product Type Probes (Labeled DNA)
Label or Dye 32P (Phosphorous-32)
Includes Label or Dye No
Labeling Target DNA (General)
Labeling Method Direct Labeling
Shipping Condition Dry Ice
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.