missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Random Primers DNA Labeling System

Product Code. 10337872
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10337872 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10337872 Supplier Invitrogen™ Supplier No. 18187013
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact 2 Buffer [500mM Tris-HCl (pH 8.0), 100mM
  • MgCl2 , 500mM NaCl], vial of dilution buffer

Ideal for radioactively labeling DNA, particularly fragments under 1 kb

  • Yields >109 cpm/μg control DNA using [ α-32 P]-dCTP
  • 25ng of DNA in one reaction
  • Performance and Quality Testing: Incorporation of a radioactively labeled nucleotide is verified using control DNA in a random primers labeling reaction

DNA and RNA Purification and Analysis, DNA Labeling, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Nucleic Acid Labeling and Oligo Synthesis, Southern Blotting

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Labeling System
Detection Method Radioactive
Content And Storage Components of the Random Primers DNA Labeling System are listed in the table to the right. Store at -20°C.
Final Product Type Probes (Labeled DNA)
Label or Dye 32P (Phosphorous-32)
Includes Label or Dye No
Labeling Target DNA (General)
Labeling Method Direct Labeling
Shipping Condition Approved for shipment on Wet or Dry Ice
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.