missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ribonuclease H

Product Code. 10378002
Change view
Click to view available options
Quantity:
120 U
30 U
Unit Size:
120 units
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10378002 120 U 120 units
10368002 30 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10378002 Supplier Invitrogen™ Supplier No. 18021071
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Ribonuclease H containing 30 units supplied at a concentration of 2U/μL

Specifically degrades RNA strand of RNA-DNA hybrid to produce 5' phosphate-terminated oligoribonucleotides and single-stranded DNA

  • Source: Purified from E. coli expressing the E. coli RNase H gene on a plasmid
  • Performance and quality testing: Ribonuclease, nonspecific endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease
  • Unit definition: One unit hydrolyzes 1nmol of RNA in 3H-labeled poly(A)poly(dT) to acid-soluble material in 20 min. at 37°C
  • Unit reaction conditions: 20mM Tris-HCl (pH 7.5), 0.1M KCl, 10mM MgCl2 , 0.1mM DTT, 5% (w/v) sucrose, 0.5nmol 3H-labeled poly(A)poly(dT), and enzyme in 50μL for 20 min. at 37°C

Removal of mRNA during second-strand cDNA synthesis; Removal of poly(A) sequences from mRNA in the presence of oligo(dT); Oligodeoxyribonucleotide-directed cleavage of RNA; PCR & Real-Time PCR; Reverse Transcription

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage 4 vials of Ribonuclease H with each containing 30 units supplied at a concentration of 2U/μl

Store at -20°C in a non-frost-free freezer.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme RNase
Quantity 120 U
Product Type Ribonuclease H

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.