Learn More
Invitrogen™ S.N.A.P.™ Gel Purification Kit
Description
Includes
Each S.N.A.P. Gel Purification Kit includes enough S.N.A.P. columns, collection vials, and reagents for 25 gel purifications.
The S.N.A.P.™ Gel Purification Kit is designed for rapid purification of DNA fragments from agarose gels. The key is unique S.N.A.P. resin, which is provided ready to use in a spin-column format. The resin efficiently binds nucleic acids up to 20 kb and eliminates organic extractions, and need to perform time-consuming ethanol precipitations.
- S.N.A.P.™ Gel Purification Kit yields pure DNA in an easy-to-perform, rapid procedure
- Excise DNA band from your agarose gel
- Add NaI solution (provided) and melt the agarose at 42–50°C
- Load sample onto the S.N.A.P.™ spin column
- Wash away contaminants
- Elute DNA in water
Specifications
Specifications
| Column Type | Spin Column |
| Content And Storage | Each S.N.A.P.™ Gel Purification Kit includes enough S.N.A.P.™ columns, collection vials, and reagents for 25 gel purifications. Store all components at room temperature. All reagents are guaranteed stable for 6 months when properly stored. |
| Format | Kit |
| Isolation Technology | Spin Column |
| Sample Type | Gel |
| For Use With (Application) | Cloning, PCR |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Label or Dye | Ethidium Bromide, SYBR Safe |
| No. of Reactions | 25 Preps |
| Product Line | S.N.A.P. |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.