missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Second Strand Buffer

Product Code. 10124942
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10124942 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124942 Supplier Invitrogen™ Supplier No. 10812014
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beinhaltet

Supplied at a 5X concentration.

Produce second-strand cDNA from mRNA

Second Strand Buffer is used with SuperScript™ cDNA Synthesis Systems to generate second-strand cDNA from mRNA in the first-strand reaction. It is supplied as a 5X concentrate and should be diluted 1:5 (1 part 5X Second Strand Buffer and 4 parts other components) prior to use.
Quality control This product has passed quality control assays that verify the absence of detectable levels of ribonuclease activity.

PCR and Real-Time PCR, Reverse Transcription

Bestellinformation

Approved for shipment on Wet or Dry ice

TRUSTED_SUSTAINABILITY

Spezifikation

Concentration 5X
Content And Storage Second-Strand Buffer is supplied at a 5X concentration. Store at -20°C.
Quantity 0.5 mL
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.