missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Spheroplast Kit for Yeast

Product Code. 10654803
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10654803 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10654803 Supplier Invitrogen™ Supplier No. K172001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

SOS medium, SE, SCE, 1M sorbitol, CaS, 40% PEG, CaT, sterile water, zymolyase, and 1M DTT. Sufficient reagents are provided in the Spheroplast Kit for 10 spheroplast preparations, each of which can be used for 5 transformations.

Contains qualified reagents for preparation of yeast spheroplasts

  • Reagents have been optimized for use with Pichia pastoris and are guaranteed by manufacturer to generate 70% spheroplasts

Cloning, Pichia pastoris Expression Systems, Protein Expression, Proteins, Expression, Isolation and Analysis, Transformation, Yeast Expression

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type Spheroplast Kit
Content And Storage The Spheroplast Kit contains the following reagents: SOS medium, SE, SCE, 1M sorbitol, CaS, 40% PEG, CaT, sterile water, zymolyase, and 1M DTT. Sufficient reagents are provided in the Spheroplast Kit for 10 spheroplast preparations, each of which can be used for 5 transformations. All components are guaranteed stable for 6 months when properly stored.
Target Organism Class P. pastoris
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.